Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01757491_m1
          See other CDYL GE Assays ›
          Gene Symbol:
          Cdyl
          Gene Name:
          chromodomain protein, Y-like
          Gene Aliases:
          -
          Chromosome Location:
          Chr.17: 29672925 - 29895386 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01757491_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cdyl
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001014145.1 NP_001014167.1 8 - 9 1733 68
          XM_006253839.2 XP_006253901.1 6 - 7 1923 68
          XM_006253840.2 XP_006253902.1 6 - 7 1783 68
          GenBank mRNA BC079003.1 - 8 - 9 1733 7112869 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1700001631.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cdyl

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          histone acetylation
          negative regulation of nucleic acid-templated transcription
          transcription corepressor activity
          histone acetyltransferase activity
          methylated histone binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P030989 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 141430 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 331035 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 365653 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 485574 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 11204 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 308986 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 110975 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 465502 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 764306 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 48613 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 229260 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 245064 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 938525 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 677562 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 634646 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 152485 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 335574 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 548815 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 535277 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 581761 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 686194 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 200533 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 814394 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 672774 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 907564 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 30869 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 244752 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 85173 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 792654 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 720086 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 583542 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 789265 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 23692 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 463300 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 717006 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 709006 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 288760 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 368377 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 419952 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 78291 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 333872 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 8233 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 711809 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 374883 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 882230 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 901870 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 701893 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 884151 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 885759 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 517383 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 167128 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 879140 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 263697 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 156609 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 256890 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 746438 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 78343 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 301827 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 67692 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 672353 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 616541 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 473135 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 540069 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 263837 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 38227 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 534109 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 449673 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 794924 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 337592 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 554741 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 839192 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 720515 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 437331 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 173126 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 656789 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 792820 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 12220 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 969551 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 27518 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 378314 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 804653 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 194665 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 365510 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 819314 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 160794 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 487578 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 158047 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 426651 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 750014 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 21663 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 187657 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 673409 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 955417 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 856089 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 827455 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 608658 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 770344 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 225736 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 609367 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 637096 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 434273 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 42276 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 312853 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 971519 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 264498 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 615222 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 844504 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 51047 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 816000 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 139046 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 648458 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 487040 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 588753 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 569786 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 786127 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 588561 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 458192 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 326434 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 204528 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 783547 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 760986 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 33418 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 101087 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 261995 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 728365 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 612869 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 733514 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 538356 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 362378 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 721120 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 536850 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 484844 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 914745 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 566244 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 793124 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 233101 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 884139 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 785118 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 957 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 242315 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 492181 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 648956 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 663626 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 47742 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 935294 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 963072 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 314567 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 269285 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 906822 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 5023 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 462745 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 371643 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 110089 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 794962 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 659705 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 306533 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 143514 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 764080 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 460665 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 433849 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 22309 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 164455 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 489 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 175897 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 41300 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 154989 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 37769 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 133316 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 138096 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 116631 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 70920 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 64157 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 95901 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 97818 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 48848 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 60960 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 174878 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 2510 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 94182 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 79050 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 42330 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 86368 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 104334 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 147196 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 157843 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 35867 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 161867 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 125459 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P030988 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P030988 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P030988 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P569063 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 90269 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 14751 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 78009 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 66643 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 104771 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 46230 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 70736 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 39882 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 2449 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 28337 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 94939 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 27709 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 13500 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 54354 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 70277 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 11054 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 102587 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 99589 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10798056 - -
          Affymetrix Rat Gene 1.0 ST Array 10798053 - -
          Affymetrix Rat Gene 1.0 ST Array 10798054 - -
          Affymetrix Rat Gene 1.0 ST Array 10798059 - -
          Affymetrix Rat Gene 1.0 ST Array 10798050 - -
          Affymetrix Rat Gene 1.0 ST Array 10798052 - -
          Affymetrix Rat Gene 1.0 ST Array 10798049 - -
          Affymetrix Rat Gene 1.0 ST Array 10798051 - -
          Affymetrix Rat Gene 1.0 ST Array 10798055 - -
          Affymetrix Rat Gene 1.0 ST Array 10798057 - -
          Affymetrix Rat Gene 1.0 ST Array 10798058 - -
          Affymetrix Rat Gene 1.1 ST Array 10798048 - -
          Rat Genome 230 2.0 Array 1384365_PM_at - -
          Rat Genome 230 2.0 Array 1374494_PM_at - -
          Rat Genome 230 2.0 Array 1378941_at - -
          Rat Genome 230 2.0 Array 1378941_PM_at - -
          Rat Genome 230 2.0 Array 1374494_at - -
          Rat Genome 230 2.0 Array 1384365_at - -
          Affymetrix HT Rat Focus Array 1384365_PM_at - -
          Affymetrix HT Rat Focus Array 1384365_at - -
          Affymetrix Rat Clariom S Assay PSR1700011266.rn.1 TC1700001631.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700011267.rn.1 TC1700001631.rn.1 RTA TC
          Rat Expression Set 230 1384365_at - -
          Rat Expression Set 230 1384365_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10798048 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21565791 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1364833 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-9kpsn:80/100.66.79.221:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0