Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01758087_g1
          See other DUS3L GE Assays ›
          Gene Symbol:
          Dus3l
          Gene Name:
          dihydrouridine synthase 3-like (S. cerevisiae)
          Gene Aliases:
          -
          Chromosome Location:
          Chr.9: 10289440 - 10294212 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01758087_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dus3l
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001034923.1 NP_001030095.1 12 - 13 1896 63
          XM_006244314.2 XP_006244376.1 12 - 13 2051 63
          GenBank mRNA BC105780.1 - 12 - 13 1896 7932299 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0900001460.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Dus3l

          Panther Classification:

          Molecular Function -

          RNA metabolism protein RNA processing factor nucleic acid metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          tRNA dihydrouridine synthesis
          oxidation-reduction process
          tRNA dihydrouridine synthase activity
          poly(A) RNA binding
          metal ion binding
          flavin adenine dinucleotide binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P539156 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P539156 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P539156 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P539156 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P539156 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 52688 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 18144 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10931380 - -
          Affymetrix Rat Gene 1.0 ST Array 10931373 - -
          Affymetrix Rat Gene 1.0 ST Array 10931381 - -
          Affymetrix Rat Gene 1.0 ST Array 10931371 - -
          Affymetrix Rat Gene 1.0 ST Array 10931372 - -
          Affymetrix Rat Gene 1.0 ST Array 10931378 - -
          Affymetrix Rat Gene 1.0 ST Array 10931369 - -
          Affymetrix Rat Gene 1.0 ST Array 10931370 - -
          Affymetrix Rat Gene 1.0 ST Array 10931379 - -
          Affymetrix Rat Gene 1.0 ST Array 10931377 - -
          Affymetrix Rat Gene 1.0 ST Array 10931368 - -
          Affymetrix Rat Gene 1.0 ST Array 10931374 - -
          Affymetrix Rat Gene 1.0 ST Array 10931376 - -
          Affymetrix Rat Gene 1.1 ST Array 10931367 - -
          Rat Genome 230 2.0 Array 1374979_PM_at - -
          Rat Genome 230 2.0 Array 1391053_at - -
          Rat Genome 230 2.0 Array 1391053_PM_at - -
          Rat Genome 230 2.0 Array 1374979_at - -
          Affymetrix HT Rat Focus Array 1374979_at - -
          Affymetrix HT Rat Focus Array 1374979_PM_at - -
          Affymetrix HT Rat Focus Array 1391053_at - -
          Affymetrix HT Rat Focus Array 1391053_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0900012041.rn.1 TC0900001460.rn.1 RTA TC
          Rat Expression Set 230 1374979_at - -
          Rat Expression Set 230 1374979_PM_at - -
          Rat Expression Set 230 1391053_PM_at - -
          Rat Expression Set 230 1391053_at - -
          Affymetrix Rat Gene 1.0 ST Array 10931367 - -
          Rat Genome U34 Set rc_AI102764_at - -
          Rat Genome U34 Set rc_AI177187_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21619098 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1361149 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline