Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01758208_m1
          See other ALDH5A1 GE Assays ›
          Gene Symbol:
          Aldh5a1
          Gene Name:
          aldehyde dehydrogenase 5 family, member A1
          Gene Aliases:
          Ssadh
          Chromosome Location:
          Chr.17: 42132085 - 42159413 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01758208_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Aldh5a1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022851.1 NP_074042.1 9 - 10 1386 58
          GenBank mRNA L34821.1 - 9 - 10 1265 58

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1700000614.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          dehydrogenase

          Gene Ontology Categories:

          Function(s) Process(es)

          glucose metabolic process
          acetate metabolic process
          succinate metabolic process
          glutamate metabolic process
          glutamine metabolic process
          glycerophospholipid metabolic process
          glucosylceramide metabolic process
          galactosylceramide metabolic process
          glutathione metabolic process
          central nervous system development
          gamma-aminobutyric acid metabolic process
          gamma-aminobutyric acid catabolic process
          post-embryonic development
          respiratory electron transport chain
          neurotransmitter catabolic process
          short-chain fatty acid metabolic process
          protein homotetramerization
          oxidation-reduction process
          aldehyde dehydrogenase (NAD) activity
          succinate-semialdehyde dehydrogenase (NAD+) activity
          succinate-semialdehyde dehydrogenase [NAD(P)+] activity
          carboxylic acid binding
          NAD binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P030919 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P067678 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P030919 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P067678 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P030919 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P067678 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P067678 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P030919 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P381822 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700637986 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 38499 - -
          Agilent Rat cDNA Microarray (G4105A) 700637986 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10795104 - -
          Affymetrix Rat Gene 1.0 ST Array 10795098 - -
          Affymetrix Rat Gene 1.0 ST Array 10795100 - -
          Affymetrix Rat Gene 1.0 ST Array 10795102 - -
          Affymetrix Rat Gene 1.0 ST Array 10795099 - -
          Affymetrix Rat Gene 1.0 ST Array 10795096 - -
          Affymetrix Rat Gene 1.0 ST Array 10795095 - -
          Affymetrix Rat Gene 1.0 ST Array 10795097 - -
          Affymetrix Rat Gene 1.0 ST Array 10795101 - -
          Affymetrix Rat Gene 1.0 ST Array 10795103 - -
          Affymetrix Rat Gene 1.1 ST Array 10795093 - -
          Rat Genome 230 2.0 Array 1391657_PM_at - -
          Rat Genome 230 2.0 Array 1391657_at - -
          Rat Genome 230 2.0 Array 1371062_PM_at - -
          Rat Genome 230 2.0 Array 1371062_at - -
          Affymetrix HT Rat Focus Array 1391657_PM_at - -
          Affymetrix HT Rat Focus Array 1371062_at - -
          Affymetrix HT Rat Focus Array 1391657_at - -
          Affymetrix HT Rat Focus Array 1371062_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1700004242.rn.1 TC1700000614.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700004243.rn.1 TC1700000614.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700004245.rn.1 TC1700000614.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700004244.rn.1 TC1700000614.rn.1 RTA TC
          Rat Expression Set 230 1371062_at - -
          Rat Expression Set 230 1371062_PM_at - -
          Rat Expression Set 230 1391657_PM_at - -
          Rat Expression Set 230 1391657_at - -
          Affymetrix Rat Gene 1.0 ST Array 10795093 - -
          Rat Genome U34 Set L34821_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21188599 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1372959 - -
          RatRef-12 Expression BeadChip ILMN_56609 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline