Search
Search
| Gene Symbol: Tada1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037980.2 | NP_001033069.2 | 6 - 7 | 794 | 60 | |
| GenBank mRNA | BC091380.1 | - | 586 | 7320760 | 60 | |
| BC158713.1 | - | 6 - 7 | 794 | 9035442 | 60 | |
Set Membership: |
|