Search
Search
| Gene Symbol: Ncf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100984.1 | NP_001094454.1 | 1 - 2 | 178 | 71 | |
| XM_006250013.2 | XP_006250075.1 | 1 - 2 | 391 | 71 | ||
| XM_008769664.1 | XP_008767886.1 | 1 - 2 | 388 | 71 | ||
| GenBank mRNA | JN864041.1 | - | 1 - 2 | 178 | 71 | |
| JN864042.1 | - | 1 - 2 | 178 | 71 | ||
| JN864045.1 | - | 1 - 2 | 178 | 71 | ||
| JN864046.1 | - | 1 - 2 | 178 | 71 | ||
Set Membership: |
|