Search
Search
| Gene Symbol: Ablim3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191698.1 | NP_001178627.1 | 8 - 9 | 1179 | 60 | |
| XM_008772143.1 | XP_008770365.1 | 8 - 9 | 1248 | 60 | ||
| XM_008772144.1 | XP_008770366.1 | 8 - 9 | 1248 | 60 | ||
| XM_008772145.1 | XP_008770367.1 | 8 - 9 | 1248 | 60 | ||
| XM_008772146.1 | XP_008770368.1 | 8 - 9 | 1248 | 60 | ||
| GenBank mRNA | FQ217083.1 | - | 4 - 5 | 343 | 60 | |
Set Membership: |
|