Search
Search

Tnfrsf26| Gene Symbol: Tnfrsf26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108511.1 | NP_001101981.1 | 2 - 3 | 132 | 63 | |
| XM_006230827.2 | XP_006230889.1 | 2 - 3 | 917 | 63 | ||
Set Membership: |
|