Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01759772_m1
          See other FFAR4 GE Assays ›
          Gene Symbol:
          Ffar4
          Gene Name:
          free fatty acid receptor 4
          Gene Aliases:
          Gpr120, O3far1
          Chromosome Location:
          Chr.1: 256786124 - 256804156 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01759772_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ffar4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001047088.1 NP_001040553.1 2 - 3 740 71
          GenBank mRNA AB207868.1 - 2 - 3 740 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100003564.rn.1

          Set Membership:

          Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          regulation of glucose transport
          cellular response to hormone stimulus
          negative regulation of apoptotic process
          hormone secretion
          negative regulation of cytokine secretion
          negative regulation of inflammatory response
          detection of chemical stimulus involved in sensory perception of taste
          positive regulation of ERK1 and ERK2 cascade
          response to peptide
          G-protein coupled receptor activity
          fatty acid binding
          taste receptor activity
          peptide binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P192083 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 119901 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 603655 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 475387 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 297634 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 374922 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 529464 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 25139 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 375042 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 599773 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 79583 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 29363 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 880583 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 576028 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 727345 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 641139 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 6939 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 7249 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 996 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P192083 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P192083 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P192083 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P192083 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 36279 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 30360 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 18014 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10715120 - -
          Affymetrix Rat Gene 1.0 ST Array 10715119 - -
          Affymetrix Rat Gene 1.0 ST Array 10715122 - -
          Affymetrix Rat Gene 1.1 ST Array 10715117 - -
          Affymetrix Rat Clariom S Assay PSR0100028907.rn.1 TC0100003564.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100028904.rn.1 TC0100003564.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100028906.rn.1 TC0100003564.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10715117 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21636823 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1362207 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline