Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctsw | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024242.1 | NP_001019413.1 | 9 - 10 | 1003 | 72 | |
GenBank mRNA | BC093401.1 | - | 9 - 10 | 1003 | 7389584 | 72 |
FQ227585.1 | - | 7 - 8 | 802 | 72 | ||
FQ235197.1 | - | 6 - 7 | 585 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |