Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01760394_m1
          See other NTN4 GE Assays ›
          Gene Symbol:
          Ntn4
          Gene Name:
          netrin 4
          Gene Aliases:
          RGD1565947
          Chromosome Location:
          Chr.7: 34533543 - 34649330 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01760394_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ntn4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001106780.1 NP_001100250.1 2 - 3 750 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700000519.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          extracellular matrix protein

          Gene Ontology Categories:

          Function(s) Process(es)

          neuron remodeling
          regulation of branching involved in salivary gland morphogenesis by extracellular matrix-epithelial cell signaling
          laminin-1 binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P148561 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P480852 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 636219 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 60145 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 234859 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 882778 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 514203 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 168748 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 130186 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 764133 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 762388 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 850297 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 312543 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 109650 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 10953 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 542965 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 20547 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 185040 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 745314 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 522043 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 841125 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 36584 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 60406 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 491691 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 222871 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 124984 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 205218 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 341052 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 392836 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 62443 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 409811 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 325558 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 394202 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 312404 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 564930 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 590375 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 392542 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 110049 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 397666 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 530375 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 3690 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 910608 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 579810 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 332847 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 314829 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 514976 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 26537 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 34518 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 113580 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 545306 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 700794 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 410172 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 900062 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 22928 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 395924 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 934990 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 603712 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 972199 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 900994 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 314382 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 250512 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 239477 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 427169 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 176848 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 81930 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 843059 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 397803 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 533877 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 825871 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 113487 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 664943 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 333519 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 397925 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 904885 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 31272 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 856677 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 128292 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 742479 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 65051 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 115694 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 6271 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 46748 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 105084 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 135096 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 147019 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 73364 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 45713 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 114310 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 58529 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 36275 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 6959 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 170963 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P480852 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P148561 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P148561 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P480852 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P480852 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P148561 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P480852 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 48664 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 4742 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10894941 - -
          Affymetrix Rat Gene 1.0 ST Array 10894940 - -
          Affymetrix Rat Gene 1.1 ST Array 10894939 - -
          Affymetrix Rat Clariom S Assay PSR0700004338.rn.1 TC0700000519.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700004334.rn.1 TC0700000519.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700004347.rn.1 TC0700000519.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700004343.rn.1 TC0700000519.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700004333.rn.1 TC0700000519.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700004340.rn.1 TC0700000519.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700004341.rn.1 TC0700000519.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700004344.rn.1 TC0700000519.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700004342.rn.1 TC0700000519.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700004336.rn.1 TC0700000519.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700004337.rn.1 TC0700000519.rn.1 RTA TC
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1360107 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-d59m7:80/100.66.79.221:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0