Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01760718_m1
          See other DSC2 GE Assays ›
          Gene Symbol:
          Dsc2
          Gene Name:
          desmocollin 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.18: 11826705 - 11858808 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01760718_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dsc2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001033688.1 NP_001028860.1 14 - 15 2475 60
          XM_008771935.1 XP_008770157.1 14 - 15 2539 60
          GenBank mRNA BC101864.1 - 14 - 15 2475 7939139 60

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1800001086.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Dsc2

          Panther Classification:

          Molecular Function -

          cadherin

          Gene Ontology Categories:

          Function(s) Process(es)

          homophilic cell adhesion via plasma membrane adhesion molecules
          cellular response to starvation
          ventricular cardiac muscle cell action potential
          cardiac muscle cell-cardiac muscle cell adhesion
          bundle of His cell to Purkinje myocyte communication
          regulation of heart rate by cardiac conduction
          calcium ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P19054 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 780344 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 216564 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 29652 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 100833 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 269863 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 842441 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 256028 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 271120 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 220494 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 426348 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 407035 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 523875 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 78430 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 550407 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 161024 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 425980 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 877175 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 124792 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 470889 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 117316 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 856124 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 307980 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 604729 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 13204 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 142589 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 35481 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 41572 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P19054 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P19054 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P19054 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P19054 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P555539 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 22903 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 28886 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 30309 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 36105 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10803367 - -
          Affymetrix Rat Gene 1.0 ST Array 10803376 - -
          Affymetrix Rat Gene 1.0 ST Array 10803362 - -
          Affymetrix Rat Gene 1.0 ST Array 10803366 - -
          Affymetrix Rat Gene 1.0 ST Array 10803368 - -
          Affymetrix Rat Gene 1.0 ST Array 10803375 - -
          Affymetrix Rat Gene 1.0 ST Array 10803364 - -
          Affymetrix Rat Gene 1.0 ST Array 10803369 - -
          Affymetrix Rat Gene 1.0 ST Array 10803361 - -
          Affymetrix Rat Gene 1.0 ST Array 10803371 - -
          Affymetrix Rat Gene 1.0 ST Array 10803373 - -
          Affymetrix Rat Gene 1.0 ST Array 10803372 - -
          Affymetrix Rat Gene 1.0 ST Array 10803363 - -
          Affymetrix Rat Gene 1.0 ST Array 10803365 - -
          Affymetrix Rat Gene 1.0 ST Array 10803374 - -
          Affymetrix Rat Gene 1.0 ST Array 10803360 - -
          Affymetrix Rat Gene 1.0 ST Array 10803370 - -
          Affymetrix Rat Gene 1.1 ST Array 10803359 - -
          Rat Genome 230 2.0 Array 1375936_at - -
          Rat Genome 230 2.0 Array 1375936_PM_at - -
          Affymetrix HT Rat Focus Array 1375936_PM_at - -
          Affymetrix HT Rat Focus Array 1375936_at - -
          Affymetrix Rat Clariom S Assay PSR1800007668.rn.1 TC1800001086.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800007669.rn.1 TC1800001086.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800007667.rn.1 TC1800001086.rn.1 RTA TC
          Rat Expression Set 230 1375936_PM_at - -
          Rat Expression Set 230 1375936_at - -
          Affymetrix Rat Gene 1.0 ST Array 10803359 - -
          Rat Genome U34 Set rc_AA851963_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21160840 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1370399 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-h9fps:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline