Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01760829_m1
          See other GFRA3 GE Assays ›
          Gene Symbol:
          Gfra3
          Gene Name:
          GDNF family receptor alpha 3
          Gene Aliases:
          -
          Chromosome Location:
          Chr.18: 27491860 - 27520133 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01760829_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gfra3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053398.1 NP_445850.1 4 - 5 831 71
          GenBank mRNA BC079378.1 - 4 - 5 831 7110848 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1800001228.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Protein Kinase Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gfra3

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          neuron migration
          signal transduction
          axon guidance
          sympathetic nervous system development
          neuron development
          receptor activity
          axon guidance receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P007618 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 7635 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 155943 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 766578 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 246061 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 347908 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 510911 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 188744 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 52820 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 283180 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 48202 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 847034 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 750715 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 283318 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 313467 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 621799 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 967994 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 461462 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 628784 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 491564 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 386598 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 145892 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 65728 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 137151 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 158915 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 147678 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P007618 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P007618 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P007618 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P151675 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P818323 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701897559 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700540944 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 32377 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 5863 - -
          Agilent Rat cDNA Microarray (G4105A) 701897559 - -
          Agilent Rat cDNA Microarray (G4105A) 700540944 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10803821 - -
          Affymetrix Rat Gene 1.0 ST Array 10803819 - -
          Affymetrix Rat Gene 1.0 ST Array 10803817 - -
          Affymetrix Rat Gene 1.0 ST Array 10803820 - -
          Affymetrix Rat Gene 1.0 ST Array 10803816 - -
          Affymetrix Rat Gene 1.0 ST Array 10803822 - -
          Affymetrix Rat Gene 1.0 ST Array 10803823 - -
          Affymetrix Rat Gene 1.0 ST Array 10803818 - -
          Affymetrix Rat Gene 1.1 ST Array 10803815 - -
          Rat Genome 230 2.0 Array 1377680_at - -
          Rat Genome 230 2.0 Array 1377680_PM_at - -
          Affymetrix HT Rat Focus Array 1377680_at - -
          Affymetrix HT Rat Focus Array 1377680_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1800008708.rn.1 TC1800001228.rn.1 RTA TC
          Rat Expression Set 230 1377680_at - -
          Rat Expression Set 230 1377680_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10803815 - -
          Rat Genome U34 Set rc_AI179473_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22118453 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1374211 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline