Search
Search
| Gene Symbol: Amelx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271073.1 | NP_001258002.1 | 2 - 3 | 124 | 111 | |
| NM_001271074.1 | NP_001258003.1 | 2 - 3 | 124 | 111 | ||
| NM_001271075.1 | NP_001258004.1 | 2 - 3 | 124 | 111 | ||
| NM_001271076.1 | NP_001258005.1 | 2 - 3 | 124 | 111 | ||
| NM_001271077.1 | NP_001258006.1 | 2 - 3 | 124 | 111 | ||
| NM_001271078.1 | NP_001258007.1 | 2 - 3 | 124 | 111 | ||
| NM_019154.1 | NP_062027.1 | 1 - 2 | 73 | 153 | ||
| GenBank mRNA | U01245.2 | - | 1 - 2 | 83 | 111 | |
| U07054.1 | - | 1 - 2 | 65 | 111 | ||
| U51195.1 | - | 2 - 3 | 125 | 111 | ||
| U60562.1 | - | 2 - 3 | 125 | 111 | ||
| U60564.1 | - | 2 - 3 | 120 | 111 | ||
| U67130.1 | - | 1 - 2 | 73 | 153 | ||
Set Membership: |
|