Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01761190_g1
          See other TNP2 GE Assays ›
          Gene Symbol:
          Tnp2
          Gene Name:
          transition protein 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 4953898 - 4954626 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01761190_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tnp2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_017057.2 NP_058753.2 1 - 2 380 67
          GenBank mRNA BC078849.1 - 1 - 2 380 7131008 67
          X14776.1 - 1 - 2 368 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000000066.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tnp2

          Panther Classification:

          Molecular Function -

          chromatin/chromatin-binding, or -regulatory protein

          Gene Ontology Categories:

          Function(s) Process(es)

          multicellular organism development
          spermatogenesis
          binding of sperm to zona pellucida
          acrosome reaction
          penetration of zona pellucida
          cell differentiation
          sperm motility
          DNA binding
          zinc ion binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P094915 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 954991 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 845104 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 570478 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 299425 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 748040 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 231835 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 146533 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 444947 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 475342 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 510472 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 739586 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 495890 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 184844 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 852065 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 364196 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 356216 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 234765 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 899462 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 170420 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 186572 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 896107 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 438635 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 756718 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 948182 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 352419 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 254505 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 771141 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 95246 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 426259 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 445283 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 947142 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 336061 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 787826 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 197413 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 280675 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 63772 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 883442 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 578123 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 550562 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 407605 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 86355 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 807724 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 323019 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 92881 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 121966 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 658493 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 71632 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 834384 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 932639 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 426405 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 421508 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 853851 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 620812 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 452743 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 337956 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 855395 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 43339 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 77201 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 710587 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 66077 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 918754 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 825687 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 287630 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 18152 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 562270 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 80009 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 861 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 698301 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 765040 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 365736 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 540801 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 118660 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 758687 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 647427 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 514930 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 923780 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 866462 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 91329 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 184224 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 701810 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 959852 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 771869 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 131985 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 42822 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 912642 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 668926 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 442840 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 747270 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 87144 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 164405 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 922169 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 324400 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 331122 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 137551 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 970165 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 681054 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 180117 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 624931 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 212975 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 900139 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 24231 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 623277 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 655328 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 924766 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 46266 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 817913 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 666561 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 879555 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 229307 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 159470 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 810910 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 559075 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 19214 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 535568 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 330120 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 15517 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 267968 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 932074 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 751504 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 112244 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 691520 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 939919 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 348852 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 676654 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 76762 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 393066 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 337253 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 822562 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 154933 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 681436 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 142493 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 134932 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 68342 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 124853 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 81045 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 75269 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 173094 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 53102 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 3475 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 7387 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 85206 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 35320 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 41716 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 150832 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 21442 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 168617 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 84391 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 25360 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 40378 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 148607 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 53301 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 147634 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 101437 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 166827 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 132578 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 56494 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 128140 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 133299 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 148359 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 59703 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 140256 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 87001 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P094915 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P094915 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P094915 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P130491 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700174656 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 84751 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 72922 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 45044 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 98146 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 54229 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 66002 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 33024 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 18936 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 87094 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 27549 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 706 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 27614 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 85375 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 92961 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 57528 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 51212 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 57220 - -
          Agilent Rat cDNA Microarray (G4105A) 700174656 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10731509 - -
          Affymetrix Rat Gene 1.0 ST Array 10731511 - -
          Affymetrix Rat Gene 1.0 ST Array 10731510 - -
          Affymetrix Rat Gene 1.1 ST Array 10731508 - -
          Rat Genome 230 2.0 Array 1370022_PM_at - -
          Rat Genome 230 2.0 Array 1370022_at - -
          Affymetrix HT Rat Focus Array 1370022_at - -
          Affymetrix HT Rat Focus Array 1370022_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1000000668.rn.1 TC1000000066.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000000669.rn.1 TC1000000066.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000000670.rn.1 TC1000000066.rn.1 RTA TC
          Rat Expression Set 230 1370022_PM_at - -
          Rat Expression Set 230 1370022_at - -
          Affymetrix Rat Gene 1.0 ST Array 10731508 - -
          Rat Genome U34 Set X14776_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22323197 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1362905 - -
          RatRef-12 Expression BeadChip ILMN_52338 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-8ltcg:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline