Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01761197_m1
          See other DEXI GE Assays ›
          Gene Symbol:
          Dexi
          Gene Name:
          dexamethasone-induced transcript
          Gene Aliases:
          RGD1564938
          Chromosome Location:
          Chr.10: 5199226 - 5211913 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01761197_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dexi
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001109026.1 NP_001102496.1 1 - 2 807 108

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000000072.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P761933 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 768805 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 370111 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 614470 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 864990 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 565986 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 147347 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 210912 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 766507 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 71903 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 178592 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 13841 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P761933 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P761933 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P761933 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 102397 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10731515 - -
          Affymetrix Rat Gene 1.0 ST Array 10731516 - -
          Affymetrix Rat Gene 1.1 ST Array 10731514 - -
          Rat Genome 230 2.0 Array 1371877_at - -
          Rat Genome 230 2.0 Array 1371877_PM_at - -
          Affymetrix HT Rat Focus Array 1371877_at - -
          Affymetrix HT Rat Focus Array 1371877_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1000000692.rn.1 TC1000000072.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000000695.rn.1 TC1000000072.rn.1 RTA TC
          Rat Expression Set 230 1371877_at - -
          Rat Expression Set 230 1371877_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10731514 - -
          Rat Genome U34 Set rc_AI101695_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21616259 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1354598 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline