Search
Search
| Gene Symbol: Snx29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109526.2 | NP_001102996.2 | 16 - 17 | 1945 | 66 | |
| XM_006245736.2 | XP_006245798.1 | 15 - 16 | 2251 | 66 | ||
| XM_008767497.1 | XP_008765719.1 | 16 - 17 | 3060 | 66 | ||
| XM_008767498.1 | XP_008765720.1 | 14 - 15 | 1886 | 66 | ||
| XM_008767499.1 | XP_008765721.1 | 9 - 10 | 3463 | 66 | ||
Set Membership: |
|