Search
Search
| Gene Symbol: Commd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109503.1 | NP_001102973.1 | 3 - 4 | 288 | 138 | |
| GenBank mRNA | BC098743.1 | - | 2 - 3 | 857 | 138 | |
| FQ229674.1 | - | 3 - 4 | 232 | 138 | ||
Set Membership: |
|