Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01761361_m1
          See other RAB33B GE Assays ›
          Gene Symbol:
          Rab33b
          Gene Name:
          RAB33B, member RAS oncogene family
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 140541619 - 140552220 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01761361_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rab33b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001108944.1 NP_001102414.1 1 - 2 659 61

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200001111.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          protein targeting to Golgi
          regulation of cell growth
          intracellular protein transport
          intra-Golgi vesicle-mediated transport
          biological_process
          metabolic process
          Rab protein signal transduction
          skeletal system morphogenesis
          regulation of epithelial cell proliferation
          regulation of Golgi organization
          negative regulation of constitutive secretory pathway
          regulation of retrograde vesicle-mediated transport, Golgi to ER
          regulation of autophagosome assembly
          molecular_function
          GTPase activity
          GTP binding
          GDP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P040635 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P573191 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 221723 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 465265 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 667983 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 899494 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 205238 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 133898 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 99987 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 182503 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 130954 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P573191 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P040635 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P040635 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P573191 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P573191 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P040635 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P328421 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10815307 - -
          Affymetrix Rat Gene 1.0 ST Array 10815306 - -
          Affymetrix Rat Gene 1.1 ST Array 10815305 - -
          Rat Genome 230 2.0 Array 1394665_at - -
          Rat Genome 230 2.0 Array 1394665_PM_at - -
          Rat Genome 230 2.0 Array 1383232_at - -
          Rat Genome 230 2.0 Array 1383232_PM_at - -
          Affymetrix HT Rat Focus Array 1383232_PM_at - -
          Affymetrix HT Rat Focus Array 1383232_at - -
          Affymetrix Rat Clariom S Assay PSR0200007055.rn.1 TC0200001111.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200007052.rn.1 TC0200001111.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200007053.rn.1 TC0200001111.rn.1 RTA TC
          Rat Expression Set 230 1383232_at - -
          Rat Expression Set 230 1383232_PM_at - -
          Rat Expression Set 230 1394665_PM_at - -
          Rat Expression Set 230 1394665_at - -
          Affymetrix Rat Gene 1.0 ST Array 10815305 - -
          Rat Genome U34 Set rc_AI639102_at - -
          Rat Genome U34 Set rc_AI639102_g_at - -
          Rat Genome U34 Set rc_AI012761_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22131327 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1360682 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline