Search
Search

RGD1563015| Gene Symbol: RGD1563015 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134627.2 | NP_001128099.2 | 3 - 4 | 322 | 80 | |
| XM_008764055.1 | XP_008762277.1 | 3 - 4 | 491 | 80 | ||
| XM_008764056.1 | XP_008762278.1 | 4 - 5 | 1085 | 80 | ||
| XM_008764057.1 | XP_008762279.1 | 2 - 3 | 419 | 80 | ||
Set Membership: |
|