Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01761735_m1
          See other KRT82 GE Assays ›
          Gene Symbol:
          Krt82
          Gene Name:
          keratin 82, type II
          Gene Aliases:
          Kb22
          Chromosome Location:
          Chr.7: 143196282 - 143206138 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01761735_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Krt82
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001008803.1 NP_001008803.1 5 - 6 872 73

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700003941.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Krt82

          Gene Ontology Categories:

          Process(es)

          structural molecule activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P120827 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 133073 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 71992 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 557447 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 802785 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 854298 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 483963 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 946578 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 542202 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 32324 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P120827 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P120827 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P120827 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P537046 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 36095 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 31320 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10907396 - -
          Affymetrix Rat Gene 1.0 ST Array 10907400 - -
          Affymetrix Rat Gene 1.0 ST Array 10907398 - -
          Affymetrix Rat Gene 1.0 ST Array 10907401 - -
          Affymetrix Rat Gene 1.0 ST Array 10907395 - -
          Affymetrix Rat Gene 1.0 ST Array 10907402 - -
          Affymetrix Rat Gene 1.0 ST Array 10907399 - -
          Affymetrix Rat Gene 1.0 ST Array 10907397 - -
          Affymetrix Rat Gene 1.0 ST Array 10907403 - -
          Affymetrix Rat Gene 1.1 ST Array 10907394 - -
          Affymetrix Rat Clariom S Assay PSR0700031786.rn.1 TC0700003941.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10907394 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1351781 - -
          RatRef-12 Expression BeadChip ILMN_47721 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline