Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01761900_m1
          See other LMBR1L GE Assays ›
          Gene Symbol:
          Lmbr1l
          Gene Name:
          limb development membrane protein 1-like
          Gene Aliases:
          RGD1309631
          Chromosome Location:
          Chr.7: 140585512 - 140600794 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01761900_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lmbr1l
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001013950.1 NP_001013972.1 6 - 7 688 110
          XM_008765698.1 XP_008763920.1 6 - 7 735 110
          XM_008765699.1 XP_008763921.1 6 - 7 731 110
          XM_008765700.1 XP_008763922.1 4 - 5 620 110
          GenBank mRNA BC089929.1 - 6 - 7 688 7308748 110

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700003865.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Lmbr1l

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P999395 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 453452 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 209294 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 346068 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 848580 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 788882 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 526462 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 99162 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P999395 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P999395 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P999395 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P999395 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 31347 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10907072 - -
          Affymetrix Rat Gene 1.0 ST Array 10907081 - -
          Affymetrix Rat Gene 1.0 ST Array 10907073 - -
          Affymetrix Rat Gene 1.0 ST Array 10907074 - -
          Affymetrix Rat Gene 1.0 ST Array 10907080 - -
          Affymetrix Rat Gene 1.0 ST Array 10907076 - -
          Affymetrix Rat Gene 1.0 ST Array 10907078 - -
          Affymetrix Rat Gene 1.0 ST Array 10907077 - -
          Affymetrix Rat Gene 1.0 ST Array 10907075 - -
          Affymetrix Rat Gene 1.1 ST Array 10907062 - -
          Rat Genome 230 2.0 Array 1391736_PM_at - -
          Rat Genome 230 2.0 Array 1391736_at - -
          Rat Genome 230 2.0 Array 1376694_PM_at - -
          Rat Genome 230 2.0 Array 1376694_at - -
          Affymetrix HT Rat Focus Array 1391736_PM_at - -
          Affymetrix HT Rat Focus Array 1376694_PM_at - -
          Affymetrix HT Rat Focus Array 1376694_at - -
          Affymetrix HT Rat Focus Array 1391736_at - -
          Affymetrix Rat Clariom S Assay PSR0700031175.rn.1 TC0700003865.rn.1 RTA TC
          Rat Expression Set 230 1376694_PM_at - -
          Rat Expression Set 230 1376694_at - -
          Rat Expression Set 230 1391736_at - -
          Rat Expression Set 230 1391736_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10907062 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22069678 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1649911 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline