Search
Search
| Gene Symbol: Hsdl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024896.1 | NP_001020067.1 | 1 - 2 | 543 | 86 | |
| XM_006255712.2 | XP_006255774.1 | 2 - 3 | 527 | 86 | ||
| XM_006255713.2 | XP_006255775.1 | 2 - 3 | 320 | 86 | ||
| XM_006255714.2 | XP_006255776.1 | 2 - 3 | 1480 | 86 | ||
| XM_008772622.1 | XP_008770844.1 | 3 - 4 | 542 | 86 | ||
| GenBank mRNA | BC097457.1 | - | 1 - 2 | 543 | 7457192 | 86 |
Set Membership: |
|