Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01762363_m1
          See other DEF8 GE Assays ›
          Gene Symbol:
          Def8
          Gene Name:
          differentially expressed in FDCP 8 homolog (mouse)
          Gene Aliases:
          -
          Chromosome Location:
          Chr.19: 56238353 - 56259204 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01762363_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Def8
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001024774.1 NP_001019945.1 11 - 12 1328 62
          XM_006255769.2 XP_006255831.1 11 - 12 1415 62
          XM_008772618.1 XP_008770840.1 11 - 12 1404 62
          GenBank mRNA BC097376.1 - 11 - 12 1328 7444965 62

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1900000813.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Def8

          Gene Ontology Categories:

          Function(s) Process(es)

          intracellular signal transduction
          zinc ion binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P885147 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 112553 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 739612 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 492454 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 266138 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 722915 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 728718 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 437196 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 415856 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 118490 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 444979 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 867991 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 26316 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 64327 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 34235 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P885147 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P885147 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P885147 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P144964 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P885147 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 4411 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 26034 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 26652 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 64653 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10808709 - -
          Affymetrix Rat Gene 1.0 ST Array 10808715 - -
          Affymetrix Rat Gene 1.0 ST Array 10808716 - -
          Affymetrix Rat Gene 1.0 ST Array 10808711 - -
          Affymetrix Rat Gene 1.0 ST Array 10808714 - -
          Affymetrix Rat Gene 1.0 ST Array 10808718 - -
          Affymetrix Rat Gene 1.0 ST Array 10808720 - -
          Affymetrix Rat Gene 1.0 ST Array 10808717 - -
          Affymetrix Rat Gene 1.0 ST Array 10808719 - -
          Affymetrix Rat Gene 1.0 ST Array 10808712 - -
          Affymetrix Rat Gene 1.0 ST Array 10808710 - -
          Affymetrix Rat Gene 1.0 ST Array 10808713 - -
          Affymetrix Rat Gene 1.1 ST Array 10808708 - -
          Rat Genome 230 2.0 Array 1384327_at - -
          Rat Genome 230 2.0 Array 1384327_PM_at - -
          Affymetrix HT Rat Focus Array 1384327_at - -
          Affymetrix HT Rat Focus Array 1384327_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1900006658.rn.1 TC1900000813.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900006660.rn.1 TC1900000813.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900006657.rn.1 TC1900000813.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900006659.rn.1 TC1900000813.rn.1 RTA TC
          Rat Expression Set 230 1384327_at - -
          Rat Expression Set 230 1384327_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10808708 - -
          Rat Genome U34 Set rc_AI231487_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1353894 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-5xfkf:80/100.66.78.86:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline