Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01762378_m1
          See other LOC498933 GE Assays ›
          Gene Symbol:
          LOC498933
          Gene Name:
          LOC498933
          Gene Aliases:
          -
          Chromosome Location:
          Chr.19: 32344078 - 32380029 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01762378_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC498933
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001025764.1 NP_001020935.1 5 - 6 601 79
          GenBank mRNA BC098038.1 - 5 - 6 601 7456623 79

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1900000427.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene LOC498933

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P974524 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 628821 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 436640 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 851505 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 882646 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 60645 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 688584 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 602059 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 876303 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 504047 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 229175 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 919216 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 607681 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 534618 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 642149 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 856039 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 893440 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 168727 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 42972 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 225049 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 24226 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 925811 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 197489 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 23319 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 64602 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 135036 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 121275 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 61318 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 125405 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P974524 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P974524 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P974524 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P974524 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 25082 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 41100 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 52629 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10807004 - -
          Affymetrix Rat Gene 1.0 ST Array 10807005 - -
          Affymetrix Rat Gene 1.0 ST Array 10807003 - -
          Affymetrix Rat Gene 1.0 ST Array 10807001 - -
          Affymetrix Rat Gene 1.0 ST Array 10807002 - -
          Affymetrix Rat Gene 1.0 ST Array 10807006 - -
          Affymetrix Rat Gene 1.1 ST Array 10807000 - -
          Affymetrix Rat Clariom S Assay PSR1900003390.rn.1 TC1900000427.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900003389.rn.1 TC1900000427.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10807000 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21429809 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1373315 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-8ltcg:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline