Search
Search
| Gene Symbol: Ces2h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044258.2 | NP_001037723.1 | 5 - 6 | 855 | 70 | |
| NM_001190346.1 | NP_001177275.1 | 5 - 6 | 855 | 70 | ||
| XM_008772523.1 | XP_008770745.1 | 5 - 6 | 874 | 70 | ||
| XM_008772524.1 | XP_008770746.1 | 6 - 7 | 997 | 70 | ||
| GenBank mRNA | BC107806.1 | - | 5 - 6 | 825 | 7368719 | 70 |
Set Membership: |
|