Search
Search

Mab21l3| Gene Symbol: Mab21l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106456.1 | NP_001099926.1 | 3 - 4 | 627 | 73 | |
| XM_006233045.2 | XP_006233107.1 | 3 - 4 | 538 | 73 | ||
| XM_006233046.2 | XP_006233108.1 | 3 - 4 | 822 | 73 | ||
| XM_008761374.1 | XP_008759596.1 | 3 - 4 | 519 | 73 | ||
| XM_008761375.1 | XP_008759597.1 | 3 - 4 | 443 | 73 | ||
Set Membership: |
|