Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01763299_g1
          See other CEP19 GE Assays ›
          Gene Symbol:
          Cep19
          Gene Name:
          centrosomal protein 19
          Gene Aliases:
          RGD621352
          Chromosome Location:
          Chr.11: 71921713 - 71938331 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01763299_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cep19
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_138865.1 NP_620220.1 3 - 4 417 79
          XM_008768696.1 XP_008766918.1 3 - 4 596 79
          XM_008768697.1 XP_008766919.1 3 - 4 582 79
          GenBank mRNA AF146738.1 - 3 - 4 417 79
          FQ211926.1 - 3 - 4 434 79

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cep19

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P480234 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 87394 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 321962 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 334109 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 154601 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 663791 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 720202 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 489090 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 54444 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P480234 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P480234 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P480234 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P480234 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701244424 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 77305 - -
          Agilent Rat cDNA Microarray (G4105A) 701244424 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10754867 - -
          Affymetrix Rat Gene 1.0 ST Array 10754869 - -
          Affymetrix Rat Gene 1.0 ST Array 10754870 - -
          Affymetrix Rat Gene 1.0 ST Array 10754868 - -
          Affymetrix Rat Gene 1.1 ST Array 10754866 - -
          Rat Genome 230 2.0 Array 1370360_PM_at - -
          Rat Genome 230 2.0 Array 1370360_at - -
          Affymetrix HT Rat Focus Array 1370360_PM_at - -
          Affymetrix HT Rat Focus Array 1370360_at - -
          Affymetrix Rat Clariom S Assay PSR1100011172.rn.1 TC1100001764.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100011170.rn.1 TC1100001764.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100011171.rn.1 TC1100001764.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100011174.rn.1 TC1100001764.rn.1 RTA TC
          Rat Expression Set 230 1370360_PM_at - -
          Rat Expression Set 230 1370360_at - -
          Affymetrix Rat Gene 1.0 ST Array 10754866 - -
          Rat Genome U34 Set rc_AI171082_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21735225 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1364181 - -
          RatRef-12 Expression BeadChip ILMN_63982 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline