Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01763381_m1
          See other SPICE1 GE Assays ›
          Gene Symbol:
          Spice1
          Gene Name:
          spindle and centriole associated protein 1
          Gene Aliases:
          Ccdc52, RGD1310729
          Chromosome Location:
          Chr.11: 61250649 - 61294120 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01763381_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Spice1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001008285.1 NP_001008286.1 16 - 17 2488 86
          XM_006248331.2 XP_006248393.1 17 - 18 2702 86
          XM_006248332.2 XP_006248394.1 16 - 17 2656 86
          XM_006248334.1 XP_006248396.1 14 - 15 2385 86
          XM_008768719.1 XP_008766941.1 16 - 17 2586 86
          XM_008768721.1 XP_008766943.1 14 - 15 2917 86
          GenBank mRNA BC085954.1 - 16 - 17 2488 7134530 86
          FQ210155.1 - 16 - 17 2553 86

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1100001589.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Spice1

          Gene Ontology Categories:

          Function(s)

          regulation of centriole replication
          cell division
          metaphase plate congression
          mitotic spindle assembly

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P840460 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 904716 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 858610 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 889704 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 956822 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 602660 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 560520 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 971035 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 584499 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 3505 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 546636 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 358121 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 565379 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 654259 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 467411 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 178992 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 126349 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 399054 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 251186 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 637784 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 546371 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 279628 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 674946 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 415621 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 49036 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 406254 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 763854 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 312023 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 329931 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 54586 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 582090 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 912855 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 135218 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 112054 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 13118 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 73498 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 5358 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P840460 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P840460 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P840460 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P840460 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 3398 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 4336 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 63691 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10754073 - -
          Affymetrix Rat Gene 1.0 ST Array 10754074 - -
          Affymetrix Rat Gene 1.0 ST Array 10754068 - -
          Affymetrix Rat Gene 1.0 ST Array 10754071 - -
          Affymetrix Rat Gene 1.0 ST Array 10754060 - -
          Affymetrix Rat Gene 1.0 ST Array 10754072 - -
          Affymetrix Rat Gene 1.0 ST Array 10754063 - -
          Affymetrix Rat Gene 1.0 ST Array 10754064 - -
          Affymetrix Rat Gene 1.0 ST Array 10754059 - -
          Affymetrix Rat Gene 1.0 ST Array 10754067 - -
          Affymetrix Rat Gene 1.0 ST Array 10754065 - -
          Affymetrix Rat Gene 1.0 ST Array 10754066 - -
          Affymetrix Rat Gene 1.0 ST Array 10754061 - -
          Affymetrix Rat Gene 1.0 ST Array 10754062 - -
          Affymetrix Rat Gene 1.0 ST Array 10754070 - -
          Affymetrix Rat Gene 1.0 ST Array 10754075 - -
          Affymetrix Rat Gene 1.0 ST Array 10754069 - -
          Affymetrix Rat Gene 1.0 ST Array 10754076 - -
          Affymetrix Rat Gene 1.1 ST Array 10754058 - -
          Rat Genome 230 2.0 Array 1391545_PM_at - -
          Rat Genome 230 2.0 Array 1391545_at - -
          Affymetrix HT Rat Focus Array 1391545_at - -
          Affymetrix HT Rat Focus Array 1391545_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1100009704.rn.1 TC1100001589.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100009706.rn.1 TC1100001589.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100009702.rn.1 TC1100001589.rn.1 RTA TC
          Rat Expression Set 230 1391545_PM_at - -
          Rat Expression Set 230 1391545_at - -
          Affymetrix Rat Gene 1.0 ST Array 10754058 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21723584 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1357257 - -
          RatRef-12 Expression BeadChip ILMN_54044 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline