Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01763512_m1
          See other DIRC2 GE Assays ›
          Gene Symbol:
          Dirc2
          Gene Name:
          disrupted in renal carcinoma 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.11: 68198709 - 68272766 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01763512_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dirc2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001012017.1 NP_001012017.1 8 - 9 1570 107
          GenBank mRNA BC081960.1 - 8 - 9 1570 7124589 107

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1100000703.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Transporter 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Dirc2

          Panther Classification:

          Molecular Function -

          secondary carrier transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          transmembrane transport
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P832637 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 395981 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 969645 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 473193 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 876984 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 159273 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 877351 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 673985 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 907849 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 284694 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 415817 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 478335 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 158868 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 811342 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 136040 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 215708 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 900277 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 233511 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 5035 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 650934 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 484991 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 442494 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 356609 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 476084 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 454938 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 152736 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 688017 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 864723 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 355966 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 402777 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 558350 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 169811 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 272319 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 733455 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 272982 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 337214 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 404884 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 572501 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 118508 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 539411 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 607794 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 748426 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 234016 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 136174 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 594531 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 81595 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 753847 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 462814 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 484045 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 115666 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 8397 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 20349 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 74201 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 118035 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 42191 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 47997 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 141850 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P832637 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P832637 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P832637 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P232290 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P832637 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 102870 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 33597 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 8650 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 58235 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 76695 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 23655 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10751493 - -
          Affymetrix Rat Gene 1.0 ST Array 10751496 - -
          Affymetrix Rat Gene 1.0 ST Array 10751489 - -
          Affymetrix Rat Gene 1.0 ST Array 10751495 - -
          Affymetrix Rat Gene 1.0 ST Array 10751491 - -
          Affymetrix Rat Gene 1.0 ST Array 10751494 - -
          Affymetrix Rat Gene 1.0 ST Array 10751490 - -
          Affymetrix Rat Gene 1.0 ST Array 10751492 - -
          Affymetrix Rat Gene 1.0 ST Array 10751497 - -
          Affymetrix Rat Gene 1.1 ST Array 10751488 - -
          Rat Genome 230 2.0 Array 1390102_at - -
          Rat Genome 230 2.0 Array 1390102_PM_at - -
          Affymetrix HT Rat Focus Array 1390102_at - -
          Affymetrix HT Rat Focus Array 1390102_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1100003830.rn.1 TC1100000703.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100003829.rn.1 TC1100000703.rn.1 RTA TC
          Rat Expression Set 230 1390102_PM_at - -
          Rat Expression Set 230 1390102_at - -
          Affymetrix Rat Gene 1.0 ST Array 10751488 - -
          Rat Genome U34 Set rc_AI168996_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376332 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline