Search
Search
| Gene Symbol: Myh14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100690.1 | NP_001094160.1 | 35 - 36 | 5057 | 53 | |
| XM_006229056.2 | XP_006229118.1 | 36 - 37 | 5257 | 53 | ||
| XM_006229057.2 | XP_006229119.1 | 35 - 36 | 5233 | 53 | ||
| GenBank mRNA | CB774028.1 | - | 2 - 3 | 339 | 53 | |
Set Membership: |
|