Search
Search

LOC102547059| Gene Symbol: LOC102547059 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006228753.2 | XP_006228815.1 | 3 - 4 | 1181 | 63 | |
| XM_006228754.2 | XP_006228816.1 | 3 - 4 | 1181 | 63 | ||
| XM_008759228.1 | XP_008757450.1 | 2 - 3 | 387 | 63 | ||
Set Membership: |
|