Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01764319_m1
          See other CEBPG GE Assays ›
          Gene Symbol:
          Cebpg
          Gene Name:
          CCAAT/enhancer binding protein (C/EBP), gamma
          Gene Aliases:
          C/EBP, CEBPRNA
          Chromosome Location:
          Chr.1: 91286955 - 91297459 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01764319_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cebpg
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012831.1 NP_036963.1 1 - 2 159 87
          XM_008759112.1 XP_008757334.1 2 - 3 402 87
          GenBank mRNA BC166589.1 - 1 - 2 148 7934938 87
          X64403.1 - 1 - 2 159 87

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the BC166589.1, NM_012831.1 transcripts.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          basic leucine zipper transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          liver development
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          immune response
          mRNA metabolic process
          B cell differentiation
          natural killer cell mediated cytotoxicity
          enucleate erythrocyte differentiation
          negative regulation of sequence-specific DNA binding transcription factor activity
          positive regulation of interferon-gamma biosynthetic process
          positive regulation of DNA repair
          positive regulation of transcription from RNA polymerase II promoter
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          DNA binding
          double-stranded DNA binding
          transcription factor activity, sequence-specific DNA binding
          protein binding
          transcription factor binding
          sequence-specific DNA binding
          protein heterodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P025098 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 342977 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 353089 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 724649 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 912691 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 212169 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 593612 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 172479 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P025098 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P025098 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P025098 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1030594 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P299451 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701573751 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 26653 - -
          Agilent Rat cDNA Microarray (G4105A) 701573751 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10721051 - -
          Affymetrix Rat Gene 1.1 ST Array 10721050 - -
          Rat Genome 230 2.0 Array 1371038_PM_at - -
          Rat Genome 230 2.0 Array 1372885_at - -
          Rat Genome 230 2.0 Array 1372885_PM_at - -
          Rat Genome 230 2.0 Array 1371038_at - -
          Affymetrix HT Rat Focus Array 1372885_PM_at - -
          Affymetrix HT Rat Focus Array 1371038_at - -
          Affymetrix HT Rat Focus Array 1371038_PM_at - -
          Affymetrix HT Rat Focus Array 1372885_at - -
          Affymetrix Rat Clariom S Assay PSR0100042254.rn.1 TC0100005222.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100042253.rn.1 TC0100005222.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100042256.rn.1 TC0100005222.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100042258.rn.1 TC0100005222.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100042255.rn.1 TC0100005222.rn.1 RTA TC
          Rat Expression Set 230 1372885_PM_at - -
          Rat Expression Set 230 1371038_PM_at - -
          Rat Expression Set 230 1371038_at - -
          Rat Expression Set 230 1372885_at - -
          Affymetrix Rat Gene 1.0 ST Array 10721050 - -
          Rat Genome U34 Set X64403_at - -
          Rat Genome U34 Set rc_AA818253_at - -
          Rat Genome U34 Set X64403_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21883983 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1370451 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline