Search
Search

Zfp74
LOC103690038| Gene Symbol: Zfp74 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271339.1 | NP_001258268.1 | 2 - 3 | 260 | 77 | |
| XM_006228731.2 | XP_006228793.1 | 3 - 4 | 1409 | 77 | ||
| XM_006228732.2 | XP_006228794.1 | 3 - 4 | 1404 | 77 | ||
| Gene Symbol: LOC103690038 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006228711.2 | XP_006228773.1 | 3 - 4 | 1409 | 77 | |
| XM_006228712.2 | XP_006228774.1 | 3 - 4 | 1404 | 77 | ||
Set Membership: |
|