Search
Search

LOC687295| Gene Symbol: LOC687295 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003748806.3 | XP_003748854.1 | 4 - 5 | 356 | 62 | |
| GenBank mRNA | FQ213435.1 | - | 3 - 4 | 117 | 62 | |
| FQ213942.1 | - | 3 - 4 | 117 | 62 | ||
| FQ216794.1 | - | 3 - 4 | 117 | 62 | ||
Set Membership: |
|