Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Rn01764643_m1
See other B4GALT1 GE Assays ›
Gene Symbol:
B4galt1
Gene Name:
UDP-Gal:betaGlcNAc beta 1,4- galactosyltransferase, polypeptide 1
Gene Aliases:
Ggtb2
Chromosome Location:
Chr.5: 57121768 - 57168610 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID
Rn01764643_m1
Best Coverage
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 250 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Inventoried
Catalog #
4331182
Price
284.00
Your Price
Online offer:
266.65
266.65
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: B4galt1
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_053287.1
NP_445739.1
2 - 3
874
72
GenBank mRNA
AA956551.1
-
2 - 3
410
72
Back To Top
More Information
Updated Mapping Information
This assay targets the same location as RTA transcript cluster(s) TC0500002556.rn.1
Set Membership:
Amplicon length between 71 and 85
Recommended Assay
Rattus norvegicus
Inventoried
Probe spans exons
Panther Classification:
Molecular Function -
glycosyltransferase
Gene Ontology Categories:
Function(s)
Process(es)
epithelial cell development
acute inflammatory response
lactose biosynthetic process
galactose metabolic process
protein glycosylation
protein N-linked glycosylation
cell adhesion
binding of sperm to zona pellucida
penetration of zona pellucida
negative regulation of cell proliferation
glycoprotein biosynthetic process
oligosaccharide biosynthetic process
response to wounding
extracellular matrix organization
mammary gland development
wound healing
regulation of cell proliferation
positive regulation of apoptotic process
development of secondary sexual characteristics
branching morphogenesis of an epithelial tube
leukocyte migration
regulation of cellular component movement
regulation of acrosome reaction
positive regulation of epithelial cell proliferation involved in wound healing
angiogenesis involved in wound healing
positive regulation of apoptotic process involved in mammary gland involution
molecular_function
beta-N-acetylglucosaminylglycopeptide beta-1,4-galactosyltransferase activity
N-acetyllactosamine synthase activity
lactose synthase activity
cytoskeletal protein binding
galactosyltransferase activity
protein kinase binding
manganese ion binding
UDP-galactosyltransferase activity
protein homodimerization activity
alpha-tubulin binding
metal ion binding
beta-tubulin binding
MicroArray Probes:
Affymetrix
Array Name
Probe IDs
Targets
Target Type
Affymetrix Rat Clariom S Assay
PSR0500018778.rn.1
TC0500002556.rn.1
RTA TC
Affymetrix Rat Clariom S Assay
PSR0500018776.rn.1
TC0500002556.rn.1
RTA TC
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.45.0-Offline