Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01764670_m1
          See other TOPORS GE Assays ›
          Gene Symbol:
          Topors
          Gene Name:
          topoisomerase I binding, arginine/serine-rich, E3 ubiquitin protein ligase
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 56554956 - 56566503 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01764670_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Topors
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001108658.1 NP_001102128.1 2 - 3 382 115

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500002546.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          ubiquitin-protein ligase

          Gene Ontology Categories:

          Function(s) Process(es)

          ubiquitin-dependent protein catabolic process
          protein monoubiquitination
          cellular response to DNA damage stimulus
          intrinsic apoptotic signaling pathway in response to DNA damage
          protein sumoylation
          protein localization to nucleus
          regulation of cell proliferation
          intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
          proteasome-mediated ubiquitin-dependent protein catabolic process
          positive regulation of transcription, DNA-templated
          positive regulation of ubiquitin-protein transferase activity
          maintenance of protein location in nucleus
          protein K48-linked ubiquitination
          DNA binding
          antigen binding
          ubiquitin-protein transferase activity
          zinc ion binding
          isomerase activity
          SUMO transferase activity
          DNA topoisomerase binding
          ubiquitin protein ligase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P012208 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 834241 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 816947 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 136784 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 232655 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 239725 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 521555 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 932997 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 115720 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 167976 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P012208 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P012208 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P012208 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P311892 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10876010 - -
          Affymetrix Rat Gene 1.0 ST Array 10876009 - -
          Affymetrix Rat Gene 1.0 ST Array 10876008 - -
          Affymetrix Rat Gene 1.1 ST Array 10876007 - -
          Rat Genome 230 2.0 Array 1392610_PM_at - -
          Rat Genome 230 2.0 Array 1392610_at - -
          Affymetrix HT Rat Focus Array 1392610_PM_at - -
          Affymetrix HT Rat Focus Array 1392610_at - -
          Affymetrix Rat Clariom S Assay PSR0500018701.rn.1 TC0500002546.rn.1 RTA TC
          Rat Expression Set 230 1392610_PM_at - -
          Rat Expression Set 230 1392610_at - -
          Affymetrix Rat Gene 1.0 ST Array 10876007 - -
          Rat Genome U34 Set rc_AI011315_at - -
          Rat Genome U34 Set rc_AI236444_at - -
          Rat Genome U34 Set rc_AI176588_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22288907 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1371948 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-j77tp:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline