Search
Search
| Gene Symbol: Cnpy4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108852.2 | NP_001102322.1 | 1 - 2 | 143 | 65 | |
| GenBank mRNA | BC162041.1 | - | 1 - 2 | 143 | 7100495 | 65 |
| FQ226241.1 | - | 1 - 2 | 162 | 65 | ||
| FQ229096.1 | - | 1 - 2 | 188 | 65 | ||
Set Membership: |
|