Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01765519_m1
          See other CPNE9 GE Assays ›
          Gene Symbol:
          Cpne9
          Gene Name:
          copine family member IX
          Gene Aliases:
          RGD1309212
          Chromosome Location:
          Chr.4: 145237967 - 145262444 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01765519_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cpne9
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001024982.1 NP_001020153.1 20 - 21 1563 97
          XM_006237022.2 XP_006237084.1 19 - 20 2440 97
          XM_008763190.1 XP_008761412.1 19 - 20 2540 97
          GenBank mRNA BC091347.1 - 20 - 21 1563 7316601 97
          FQ213359.1 - 11 - 12 816 97

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400001566.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cpne9

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P468231 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 15826 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 828386 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 532575 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 476723 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 747328 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 421002 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 447934 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 589721 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 247833 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 795182 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 375623 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 475220 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 437717 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 234054 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 606971 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 406607 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 311695 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 158077 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 243 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P468231 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P468231 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P468231 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P468231 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 42677 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 62076 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10857720 - -
          Affymetrix Rat Gene 1.0 ST Array 10857737 - -
          Affymetrix Rat Gene 1.0 ST Array 10857732 - -
          Affymetrix Rat Gene 1.0 ST Array 10857725 - -
          Affymetrix Rat Gene 1.0 ST Array 10857736 - -
          Affymetrix Rat Gene 1.0 ST Array 10857722 - -
          Affymetrix Rat Gene 1.0 ST Array 10857724 - -
          Affymetrix Rat Gene 1.0 ST Array 10857733 - -
          Affymetrix Rat Gene 1.0 ST Array 10857729 - -
          Affymetrix Rat Gene 1.0 ST Array 10857734 - -
          Affymetrix Rat Gene 1.0 ST Array 10857735 - -
          Affymetrix Rat Gene 1.0 ST Array 10857730 - -
          Affymetrix Rat Gene 1.0 ST Array 10857738 - -
          Affymetrix Rat Gene 1.0 ST Array 10857728 - -
          Affymetrix Rat Gene 1.0 ST Array 10857723 - -
          Affymetrix Rat Gene 1.0 ST Array 10857731 - -
          Affymetrix Rat Gene 1.0 ST Array 10857727 - -
          Affymetrix Rat Gene 1.0 ST Array 10857726 - -
          Affymetrix Rat Gene 1.0 ST Array 10857721 - -
          Affymetrix Rat Gene 1.1 ST Array 10857718 - -
          Rat Genome 230 2.0 Array 1377168_PM_at - -
          Rat Genome 230 2.0 Array 1377168_at - -
          Affymetrix HT Rat Focus Array 1377168_at - -
          Affymetrix HT Rat Focus Array 1377168_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0400011007.rn.1 TC0400001566.rn.1 RTA TC
          Rat Expression Set 230 1377168_at - -
          Rat Expression Set 230 1377168_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10857718 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1370899 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-d59m7:80/100.66.79.221:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0