Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01765626_m1
          See other EEFSEC GE Assays ›
          Gene Symbol:
          Eefsec
          Gene Name:
          eukaryotic elongation factor, selenocysteine-tRNA-specific
          Gene Aliases:
          RGD1560552
          Chromosome Location:
          Chr.4: 120194335 - 120390202 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01765626_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Eefsec
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001109249.1 NP_001102719.1 3 - 4 624 62
          XM_006236864.2 XP_006236926.1 3 - 4 638 62
          XM_008763079.1 XP_008761301.1 3 - 4 639 62
          GenBank mRNA BC168694.1 - 3 - 4 601 7937199 62

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400003291.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          translation elongation factor

          Gene Ontology Categories:

          Function(s) Process(es)

          selenocysteine incorporation
          metabolic process
          tRNA binding
          translation elongation factor activity
          GTPase activity
          GTP binding
          selenocysteine insertion sequence binding
          ribonucleoprotein complex binding

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P17852 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 840095 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 318677 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 68896 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 686797 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 25099 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 163336 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 248201 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 81720 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 75736 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 258397 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 113494 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 59303 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 535754 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 760982 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 444524 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 636976 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 827690 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 507980 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 666328 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 101973 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 640671 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 462790 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 238288 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 573587 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 706716 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 297625 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 882192 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 369085 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 395589 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 410850 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 675849 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 755878 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 699195 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 526106 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 627800 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 965533 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 410190 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 655184 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 924379 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 590968 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 698246 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 120380 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 594905 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 508113 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 264981 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 606178 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 62596 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 231444 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 389455 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 326980 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 506284 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 557065 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 38219 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 234663 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 340679 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 963107 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 577693 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 874670 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 690964 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 509682 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 256459 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 912795 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 656850 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 415905 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 701532 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 576803 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 880467 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 88765 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 119535 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 38129 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 919666 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 497 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 428346 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 848288 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 604217 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 752261 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 852102 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 254921 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 285126 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 379701 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 748788 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 321937 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 51278 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 627091 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 803698 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 683891 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 205553 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 380755 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 479103 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 190455 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 408065 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 574915 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 823547 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 838429 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 144143 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 660582 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 498858 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 835366 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 151202 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 871619 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 543241 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 764197 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 747539 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 681095 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 316031 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 390094 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 382396 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 798616 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 356394 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 920425 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 266493 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 349523 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 322169 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 929302 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 331121 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 202546 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 373152 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 564089 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 193992 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 602954 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 274892 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 459166 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 431999 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 856285 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 653142 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 580361 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 415660 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 727822 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 354904 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 523328 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 87523 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 433655 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 333474 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 118473 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 7615 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 17238 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 4820 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 965578 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 736096 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 897500 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 843141 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 607571 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 570535 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 891007 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 37930 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 634505 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 320433 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 596772 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 599545 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 98274 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 46258 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 41546 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 13249 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 59650 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 93884 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 88876 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 82471 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 7623 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 148549 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 164813 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 107395 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 70458 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 61106 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 122197 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 1914 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 70006 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 129844 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 10352 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 43762 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 73129 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 151668 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 30929 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 137806 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P17852 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P17852 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P17852 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P17852 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P983152 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 86957 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 57298 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 93759 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 57245 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 8011 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10863894 - -
          Affymetrix Rat Gene 1.0 ST Array 10863892 - -
          Affymetrix Rat Gene 1.0 ST Array 10863891 - -
          Affymetrix Rat Gene 1.0 ST Array 10863895 - -
          Affymetrix Rat Gene 1.0 ST Array 10863896 - -
          Affymetrix Rat Gene 1.0 ST Array 10863897 - -
          Affymetrix Rat Gene 1.0 ST Array 10863893 - -
          Affymetrix Rat Gene 1.1 ST Array 10863890 - -
          Rat Genome 230 2.0 Array 1383558_PM_at - -
          Rat Genome 230 2.0 Array 1383558_at - -
          Affymetrix HT Rat Focus Array 1383558_PM_at - -
          Affymetrix HT Rat Focus Array 1383558_at - -
          Affymetrix Rat Clariom S Assay PSR0400023395.rn.1 TC0400003291.rn.1 RTA TC
          Rat Expression Set 230 1383558_at - -
          Rat Expression Set 230 1383558_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10863890 - -
          Rat Genome U34 Set rc_AA964957_at - -
          Rat Genome U34 Set rc_AI072379_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20744231 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline