Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01765694_m1
          See other RNF32 GE Assays ›
          Gene Symbol:
          Rnf32
          Gene Name:
          ring finger protein 32
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 2053283 - 2113917 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01765694_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rnf32
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001012095.1 NP_001012095.1 10 - 11 1424 74
          XM_006235853.2 XP_006235915.1 8 - 9 1022 74
          XM_006235854.2 XP_006235916.1 8 - 9 1021 74
          XM_006235856.2 XP_006235918.1 9 - 10 1078 74
          GenBank mRNA BC083635.1 - 10 - 11 1424 7134691 74

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400000045.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rnf32

          Gene Ontology Categories:

          Process(es)

          zinc ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P064481 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P21153 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 520392 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 141520 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 256981 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 931677 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 832331 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 477726 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 84577 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 263815 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 304306 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 5544 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 61666 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 341675 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 397153 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 787753 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 372425 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 888242 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 633048 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 786475 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 42927 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 323803 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 362696 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 499607 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 286916 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 210082 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 152845 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 882628 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 220506 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 107697 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 625093 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 603984 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 572019 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 61653 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 56950 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 120427 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 67984 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 48052 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 48779 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 83547 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 97616 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P064481 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P21153 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P064481 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P21153 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P21153 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P064481 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P265831 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 72825 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 89379 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 45709 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 8626 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10859853 - -
          Affymetrix Rat Gene 1.0 ST Array 10859847 - -
          Affymetrix Rat Gene 1.0 ST Array 10859841 - -
          Affymetrix Rat Gene 1.0 ST Array 10859850 - -
          Affymetrix Rat Gene 1.0 ST Array 10859851 - -
          Affymetrix Rat Gene 1.0 ST Array 10859845 - -
          Affymetrix Rat Gene 1.0 ST Array 10859846 - -
          Affymetrix Rat Gene 1.0 ST Array 10859842 - -
          Affymetrix Rat Gene 1.0 ST Array 10859843 - -
          Affymetrix Rat Gene 1.0 ST Array 10859848 - -
          Affymetrix Rat Gene 1.0 ST Array 10859849 - -
          Affymetrix Rat Gene 1.0 ST Array 10859844 - -
          Affymetrix Rat Gene 1.0 ST Array 10859852 - -
          Affymetrix Rat Gene 1.1 ST Array 10859840 - -
          Rat Genome 230 2.0 Array 1391667_at - -
          Rat Genome 230 2.0 Array 1391667_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0400000205.rn.1 TC0400000045.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400000206.rn.1 TC0400000045.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400000207.rn.1 TC0400000045.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10859840 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20819104 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376392 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline