Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01765845_g1
          See other UBE2K GE Assays ›
          Gene Symbol:
          Ube2k
          Gene Name:
          ubiquitin-conjugating enzyme E2K
          Gene Aliases:
          E2-25k, Hip2, Hypg, Lig
          Chromosome Location:
          Chr.14: 44289247 - 44349119 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01765845_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ube2k
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001106006.1 NP_001099476.1 6 - 7 738 119
          XM_006250934.2 XP_006250996.1 6 - 7 935 119
          XM_008770140.1 XP_008768362.1 4 - 5 535 119
          GenBank mRNA FQ226880.1 - 6 - 7 730 119
          FQ233155.1 - 6 - 7 686 119
          FQ234076.1 - 6 - 7 716 119

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          ubiquitin-protein ligase

          Gene Ontology Categories:

          Function(s) Process(es)

          protein polyubiquitination
          regulation of proteasomal ubiquitin-dependent protein catabolic process
          proteasome-mediated ubiquitin-dependent protein catabolic process
          intrinsic apoptotic signaling pathway in response to endoplasmic reticulum stress
          protein K48-linked ubiquitination
          ubiquitin-protein transferase activity
          ligase activity
          ubiquitin-like protein transferase activity
          ubiquitin protein ligase binding
          ubiquitin-ubiquitin ligase activity
          ubiquitin protein ligase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P066003 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P573232 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 731522 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 506882 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 912759 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 349912 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 865848 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 155714 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 171131 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 330692 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 57154 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 612299 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 580304 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 472662 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 136723 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 853509 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 959169 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 636869 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 189208 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 255373 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 478240 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 568902 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 823625 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 671217 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 654707 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 425477 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 207179 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 507508 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 773642 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 470578 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 356565 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 645863 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 143125 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 417517 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 654201 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 944608 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 99908 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 122817 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 444942 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 900579 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 388067 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 116718 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 302082 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 255756 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 865445 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 54842 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 350911 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 405213 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 71724 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 254995 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 900814 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 941913 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 504924 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 546127 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 737184 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 35506 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 60910 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 66537 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 59600 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 112683 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 71053 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 166796 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 10977 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 91461 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 49538 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 136436 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P066003 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P573232 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P066003 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P573232 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P573232 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P066003 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P842057 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 9664 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 104485 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 23482 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 84369 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10776801 - -
          Affymetrix Rat Gene 1.0 ST Array 10776805 - -
          Affymetrix Rat Gene 1.0 ST Array 10776802 - -
          Affymetrix Rat Gene 1.0 ST Array 10776804 - -
          Affymetrix Rat Gene 1.0 ST Array 10776806 - -
          Affymetrix Rat Gene 1.0 ST Array 10776803 - -
          Affymetrix Rat Gene 1.1 ST Array 10776800 - -
          Rat Genome 230 2.0 Array 1385259_at - -
          Rat Genome 230 2.0 Array 1375965_PM_at - -
          Rat Genome 230 2.0 Array 1375965_at - -
          Rat Genome 230 2.0 Array 1385259_PM_at - -
          Rat Genome 230 2.0 Array 1383515_PM_at - -
          Rat Genome 230 2.0 Array 1383515_at - -
          Rat Genome 230 2.0 Array 1373063_PM_at - -
          Rat Genome 230 2.0 Array 1373063_at - -
          Affymetrix HT Rat Focus Array 1385259_PM_at - -
          Affymetrix HT Rat Focus Array 1375965_at - -
          Affymetrix HT Rat Focus Array 1373063_PM_at - -
          Affymetrix HT Rat Focus Array 1385259_at - -
          Affymetrix HT Rat Focus Array 1375965_PM_at - -
          Affymetrix HT Rat Focus Array 1373063_at - -
          Affymetrix Rat Clariom S Assay PSR1400013025.rn.1 TC1400001773.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400013029.rn.1 TC1400001773.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400013030.rn.1 TC1400001773.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400013026.rn.1 TC1400001773.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400013027.rn.1 TC1400001773.rn.1 RTA TC
          Rat Expression Set 230 1375965_at - -
          Rat Expression Set 230 1373063_PM_at - -
          Rat Expression Set 230 1373063_at - -
          Rat Expression Set 230 1375965_PM_at - -
          Rat Expression Set 230 1385259_at - -
          Rat Expression Set 230 1385259_PM_at - -
          Rat Expression Set 230 1383515_at - -
          Rat Expression Set 230 1383515_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10776800 - -
          Rat Genome U34 Set rc_AA998447_at - -
          Rat Genome U34 Set rc_AI059463_at - -
          Rat Genome U34 Set rc_AI179857_at - -
          Rat Genome U34 Set rc_AI229167_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21980381 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1370917 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-j77tp:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline