Search
Search

LOC689986| Gene Symbol: LOC689986 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109563.1 | NP_001103033.1 | 4 - 5 | 2612 | 112 | |
| XM_006250572.2 | XP_006250634.1 | 5 - 6 | 879 | 112 | ||
| XM_006250573.2 | XP_006250635.1 | 5 - 6 | 883 | 112 | ||
| XM_006250574.2 | XP_006250636.1 | 5 - 6 | 783 | 112 | ||
| XM_006250575.2 | XP_006250637.1 | 4 - 5 | 730 | 112 | ||
| XM_006250576.2 | XP_006250638.1 | 4 - 5 | 634 | 112 | ||
| XM_006250580.2 | XP_006250642.1 | 5 - 6 | 878 | 112 | ||
| XM_006250581.2 | XP_006250643.1 | 5 - 6 | 880 | 112 | ||
| XM_008769942.1 | XP_008768164.1 | 6 - 7 | 927 | 112 | ||
Set Membership: |
|