Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01766076_m1
          See other TNFAIP3 GE Assays ›
          Gene Symbol:
          Tnfaip3
          Gene Name:
          tumor necrosis factor, alpha-induced protein 3
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 14401103 - 14412822 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01766076_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tnfaip3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_003748656.3 XP_003748704.2 2 - 3 606 64

          Back To Top

          More Information


          Set Membership:

          Amplicon length less than or equal to 70 Apoptosis Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          cysteine protease

          Gene Ontology Categories:

          Function(s) Process(es)

          B-1 B cell homeostasis
          response to molecule of bacterial origin
          regulation of germinal center formation
          negative regulation of chronic inflammatory response
          protein deubiquitination
          negative regulation of protein ubiquitination
          negative regulation of NF-kappaB transcription factor activity
          response to muramyl dipeptide
          negative regulation of interleukin-1 beta production
          negative regulation of interleukin-2 production
          negative regulation of interleukin-6 production
          negative regulation of tumor necrosis factor production
          negative regulation of toll-like receptor 3 signaling pathway
          negative regulation of toll-like receptor 5 signaling pathway
          protein K11-linked deubiquitination
          negative regulation of I-kappaB kinase/NF-kappaB signaling
          positive regulation of protein catabolic process
          negative regulation of cyclin-dependent protein serine/threonine kinase activity
          negative regulation of innate immune response
          negative regulation of smooth muscle cell proliferation
          negative regulation of inflammatory response
          negative regulation of B cell activation
          negative regulation of cell death
          cellular response to hydrogen peroxide
          negative regulation of nucleotide-binding oligomerization domain containing 1 signaling pathway
          negative regulation of nucleotide-binding oligomerization domain containing 2 signaling pathway
          protein K63-linked deubiquitination
          protein K48-linked ubiquitination
          protein K48-linked deubiquitination
          cellular response to lipopolysaccharide
          tolerance induction to lipopolysaccharide
          establishment of protein localization to vacuole
          negative regulation of extrinsic apoptotic signaling pathway via death domain receptors
          positive regulation of cellular protein catabolic process
          positive regulation of hepatocyte proliferation
          negative regulation of CD40 signaling pathway
          negative regulation of endothelial cell apoptotic process
          protease binding
          DNA binding
          ubiquitin-protein transferase activity
          thiol-dependent ubiquitin-specific protease activity
          zinc ion binding
          kinase binding
          ubiquitin binding
          protein self-association

          MicroArray Probes:

          ABI

          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21878791 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-knt4q:80/100.66.75.14:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0