Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01766084_m1
          See other KATNA1 GE Assays ›
          Gene Symbol:
          Katna1
          Gene Name:
          katanin p60 (ATPase-containing) subunit A1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 1826170 - 1867786 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01766084_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Katna1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001004217.4 NP_001004217.2 10 - 11 1471 62
          GenBank mRNA AY621629.1 - 9 - 10 1275 62
          BC097929.1 - 10 - 11 1471 7444887 62
          BI302698.1 - 1 - 2 62

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100000022.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Katna1

          Panther Classification:

          Molecular Function -

          non-motor microtubule binding protein

          Gene Ontology Categories:

          Function(s) Process(es)

          microtubule bundle formation
          neuron migration
          mitotic nuclear division
          protein localization
          metabolic process
          negative regulation of neuron projection development
          protein ubiquitination
          cytoplasmic microtubule organization
          microtubule severing
          cell division
          ubiquitin-protein transferase activity
          ATP binding
          microtubule binding
          microtubule-severing ATPase activity
          ATPase activity
          dynein binding
          protein heterodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P344267 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 165281 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 2217 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 131272 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 275262 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 270431 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 470877 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 624701 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 265480 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 359864 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 218062 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 26631 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 477662 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 176002 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 330970 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 602640 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 755521 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 844537 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 78561 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 539632 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 965864 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 555309 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 165017 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 433368 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 357355 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 74528 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 257309 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 918960 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 129214 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 41169 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 80948 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 136823 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 170074 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 173446 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 53395 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P344267 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P344267 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P344267 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P344267 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 13220 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 4002 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 59797 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 77303 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10701724 - -
          Affymetrix Rat Gene 1.0 ST Array 10701725 - -
          Affymetrix Rat Gene 1.0 ST Array 10701722 - -
          Affymetrix Rat Gene 1.0 ST Array 10701723 - -
          Affymetrix Rat Gene 1.0 ST Array 10701726 - -
          Affymetrix Rat Gene 1.0 ST Array 10701721 - -
          Affymetrix Rat Gene 1.0 ST Array 10701718 - -
          Affymetrix Rat Gene 1.0 ST Array 10701720 - -
          Affymetrix Rat Gene 1.0 ST Array 10701727 - -
          Affymetrix Rat Gene 1.0 ST Array 10701719 - -
          Affymetrix Rat Gene 1.0 ST Array 10701729 - -
          Affymetrix Rat Gene 1.0 ST Array 10701728 - -
          Affymetrix Rat Gene 1.1 ST Array 10701717 - -
          Rat Genome 230 2.0 Array 1391086_at - -
          Rat Genome 230 2.0 Array 1391086_PM_at - -
          Affymetrix HT Rat Focus Array 1391086_at - -
          Affymetrix HT Rat Focus Array 1391086_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100000141.rn.1 TC0100000022.rn.1 RTA TC
          Rat Expression Set 230 1391086_at - -
          Rat Expression Set 230 1391086_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10701717 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21130075 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376655 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline