Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01766141_m1
          See other PKD2L1 GE Assays ›
          Gene Symbol:
          Pkd2l1
          Gene Name:
          polycystic kidney disease 2-like 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 263922201 - 263959318 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01766141_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pkd2l1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001106352.1 NP_001099822.1 9 - 10 1820 89
          XM_008760399.1 XP_008758621.1 9 - 10 1655 89

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100007762.rn.1

          Set Membership:

          Amplicon length between 86 and 100 Ion Channel Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          detection of chemical stimulus involved in sensory perception of sour taste
          cation transport
          smoothened signaling pathway
          sodium ion transmembrane transport
          sensory perception of sour taste
          detection of mechanical stimulus
          protein homotrimerization
          calcium ion transmembrane transport
          cellular response to acidic pH
          potassium ion transmembrane transport
          cation transmembrane transport
          calcium activated cation channel activity
          cation channel activity
          calcium channel activity
          sodium channel activity
          calcium ion binding
          cytoskeletal protein binding
          cation transmembrane transporter activity
          calcium-activated potassium channel activity
          sour taste receptor activity
          identical protein binding
          muscle alpha-actinin binding
          alpha-actinin binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P062643 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P062648 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 486598 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 857581 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 813517 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 758702 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 664217 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 128927 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 896874 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 34642 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 919637 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 776982 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 440449 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 912165 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 25592 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 430208 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 52136 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 349291 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 866620 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 797631 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 81881 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 73621 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 186799 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 509533 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 252143 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 10636 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 584567 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 1285 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 280301 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 907312 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 118207 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 71657 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 113696 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 25556 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P062643 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P062648 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P062648 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P062643 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P062643 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P062648 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P501426 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 37116 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 5940 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10715469 - -
          Affymetrix Rat Gene 1.0 ST Array 10715468 - -
          Affymetrix Rat Gene 1.0 ST Array 10715462 - -
          Affymetrix Rat Gene 1.0 ST Array 10715466 - -
          Affymetrix Rat Gene 1.0 ST Array 10715464 - -
          Affymetrix Rat Gene 1.0 ST Array 10715471 - -
          Affymetrix Rat Gene 1.0 ST Array 10715470 - -
          Affymetrix Rat Gene 1.0 ST Array 10715463 - -
          Affymetrix Rat Gene 1.0 ST Array 10715465 - -
          Affymetrix Rat Gene 1.0 ST Array 10715467 - -
          Affymetrix Rat Gene 1.0 ST Array 10715474 - -
          Affymetrix Rat Gene 1.1 ST Array 10715461 - -
          Affymetrix Rat Clariom S Assay PSR0100062568.rn.1 TC0100007762.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10715461 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22142505 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1360198 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline