Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01766338_m1
          See other DPCD GE Assays ›
          Gene Symbol:
          Dpcd
          Gene Name:
          deleted in primary ciliary dyskinesia
          Gene Aliases:
          RGD1307648
          Chromosome Location:
          Chr.1: 265298880 - 265318521 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01766338_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dpcd
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001013905.1 NP_001013927.1 2 - 3 172 85
          XM_006231450.2 XP_006231512.1 2 - 3 202 85
          GenBank mRNA BC078988.1 - 2 - 3 172 7112530 85

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the BC078988.1, NM_001013905.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Dpcd

          Gene Ontology Categories:

          Function(s)

          epithelial cilium movement
          spermatogenesis
          determination of left/right symmetry
          ventricular system development
          lateral ventricle development
          third ventricle development
          sperm motility
          left/right pattern formation

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P161413 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 626254 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 567599 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 662076 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 540447 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 338226 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 325679 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 162716 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 27968 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 389423 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 724849 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 347882 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 781513 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 829895 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 441701 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 150791 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 15893 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 20760 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P161413 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P161413 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P161413 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P10249 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 26502 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 85308 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 51229 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10715701 - -
          Affymetrix Rat Gene 1.0 ST Array 10715703 - -
          Affymetrix Rat Gene 1.0 ST Array 10715698 - -
          Affymetrix Rat Gene 1.0 ST Array 10715700 - -
          Affymetrix Rat Gene 1.0 ST Array 10715702 - -
          Affymetrix Rat Gene 1.0 ST Array 10715699 - -
          Affymetrix Rat Gene 1.1 ST Array 10715697 - -
          Rat Genome 230 2.0 Array 1374358_at - -
          Rat Genome 230 2.0 Array 1377431_PM_at - -
          Rat Genome 230 2.0 Array 1374358_PM_at - -
          Rat Genome 230 2.0 Array 1377431_at - -
          Affymetrix HT Rat Focus Array 1377431_PM_at - -
          Affymetrix HT Rat Focus Array 1374358_PM_at - -
          Affymetrix HT Rat Focus Array 1377431_at - -
          Affymetrix HT Rat Focus Array 1374358_at - -
          Affymetrix Rat Clariom S Assay PSR0100030240.rn.1 TC0100003711.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100030236.rn.1 TC0100003711.rn.1 RTA TC
          Rat Expression Set 230 1374358_PM_at - -
          Rat Expression Set 230 1374358_at - -
          Rat Expression Set 230 1377431_at - -
          Rat Expression Set 230 1377431_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10715697 - -
          Rat Genome U34 Set rc_AA956318_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21280748 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1373961 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline