Search
Search
| Gene Symbol: Scaper | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008766363.1 | XP_008764585.1 | 30 - 31 | 4092 | 56 | |
| XM_008766364.1 | XP_008764586.1 | 30 - 31 | 4197 | 56 | ||
| XM_008766365.1 | XP_008764587.1 | 30 - 31 | 4074 | 56 | ||
Set Membership: |
|