Search
Search

Ubash3b| Gene Symbol: Ubash3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191792.1 | NP_001178721.1 | 7 - 8 | 1117 | 65 | |
| XM_006242848.2 | XP_006242910.1 | 7 - 8 | 1442 | 65 | ||
| XM_006242850.2 | XP_006242912.1 | 7 - 8 | 1691 | 65 | ||
| XM_008766123.1 | XP_008764345.1 | 7 - 8 | 1445 | 65 | ||
| GenBank mRNA | FQ221553.1 | - | 7 - 8 | 1368 | 65 | |
| FQ223998.1 | - | 7 - 8 | 1368 | 65 | ||
Set Membership: |
|