Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01766950_m1
          See other ETFA GE Assays ›
          Gene Symbol:
          Etfa
          Gene Name:
          electron-transfer-flavoprotein, alpha polypeptide
          Gene Aliases:
          ETF
          Chromosome Location:
          Chr.8: 60028786 - 60086352 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01766950_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Etfa
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001009668.1 NP_001009668.1 10 - 11 962 77
          GenBank mRNA BC088412.1 - 10 - 11 962 7108329 77
          FQ219339.1 - 10 - 11 986 77

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0800002596.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Etfa

          Panther Classification:

          Molecular Function -

          oxidoreductase

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          oxidation-reduction process
          electron carrier activity
          oxidoreductase activity
          flavin adenine dinucleotide binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P18509 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 793406 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 607579 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 627123 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 197892 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 653824 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 318691 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 191281 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 789419 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 332035 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 455301 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 826978 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 850451 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 338558 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 924511 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 116984 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 700882 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 571588 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 816995 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 263390 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 281309 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 649709 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 829475 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 82165 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 30280 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 771264 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 547197 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 950515 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 100553 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 722779 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 526359 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 438182 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 833844 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 98997 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 478888 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 706220 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 940737 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 117039 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 716269 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 688535 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 111937 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 816720 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 654948 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 663567 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 885224 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 69682 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 45869 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 136344 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 136403 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 29513 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 3132 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 30650 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 152173 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P18509 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P18509 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P18509 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P18509 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 99571 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 9316 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 8080 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 28607 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10917627 - -
          Affymetrix Rat Gene 1.0 ST Array 10917623 - -
          Affymetrix Rat Gene 1.0 ST Array 10917625 - -
          Affymetrix Rat Gene 1.0 ST Array 10917630 - -
          Affymetrix Rat Gene 1.0 ST Array 10917634 - -
          Affymetrix Rat Gene 1.0 ST Array 10917632 - -
          Affymetrix Rat Gene 1.0 ST Array 10917624 - -
          Affymetrix Rat Gene 1.0 ST Array 10917631 - -
          Affymetrix Rat Gene 1.0 ST Array 10917633 - -
          Affymetrix Rat Gene 1.0 ST Array 10917628 - -
          Affymetrix Rat Gene 1.0 ST Array 10917626 - -
          Affymetrix Rat Gene 1.0 ST Array 10917629 - -
          Affymetrix Rat Gene 1.1 ST Array 10917622 - -
          Rat Genome 230 2.0 Array 1371253_at - -
          Rat Genome 230 2.0 Array 1371253_PM_at - -
          Affymetrix HT Rat Focus Array 1371253_at - -
          Affymetrix HT Rat Focus Array 1371253_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0800020967.rn.1 TC0800002596.rn.1 RTA TC
          Rat Expression Set 230 1371253_PM_at - -
          Rat Expression Set 230 1371253_at - -
          Affymetrix Rat Gene 1.0 ST Array 10917622 - -
          Rat Genome U34 Set rc_AA894174_at - -
          Rat Genome U34 Set rc_AA894174_g_at - -
          Rat Genome U34 Set rc_AI171197_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21711464 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1365910 - -
          RatRef-12 Expression BeadChip ILMN_62187 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline