Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01767026_m1
          See other CEP57L1 GE Assays ›
          Gene Symbol:
          Cep57l1
          Gene Name:
          centrosomal protein 57-like 1
          Gene Aliases:
          RGD1311358
          Chromosome Location:
          Chr.20: 46610144 - 46666818 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01767026_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cep57l1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001017448.1 NP_001017448.1 10 - 11 1194 91
          XM_006256562.2 XP_006256624.1 10 - 11 1475 91
          XM_006256563.2 XP_006256625.1 10 - 11 1430 91
          XM_006256564.2 XP_006256626.1 10 - 11 1230 91
          XM_006256565.1 XP_006256627.1 11 - 12 1253 91
          XM_006256566.2 XP_006256628.1 10 - 11 1887 91
          XM_006256567.2 XP_006256629.1 11 - 12 1222 91
          XM_006256568.2 XP_006256630.1 9 - 10 1105 91
          XM_008772982.1 XP_008771204.1 10 - 11 2145 91
          XM_008772983.1 XP_008771205.1 10 - 11 1218 91
          GenBank mRNA BC079256.1 - 10 - 11 1194 7133287 91

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC2000001708.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cep57l1

          Gene Ontology Categories:

          Function(s) Process(es)

          microtubule anchoring
          microtubule binding
          identical protein binding
          gamma-tubulin binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P267501 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 880037 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 555286 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 92914 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 812551 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 328580 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 756172 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 220926 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 399665 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 77988 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 65708 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 486718 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 425350 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 78806 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 651902 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 351159 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 499252 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 65457 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 477146 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 533151 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 101236 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 212394 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 929838 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 336895 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 391341 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 952729 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 584311 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 597648 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 407874 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 779838 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 516413 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 557802 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 437382 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 667486 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 99019 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 538483 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 712479 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 281180 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 373303 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 634874 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 3812 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 282932 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 159102 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 36475 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 62757 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 5977 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 4475 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 131339 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 64466 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 47899 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P267501 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P267501 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P267501 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P267501 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 53682 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 25760 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 90950 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 35079 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10833782 - -
          Affymetrix Rat Gene 1.0 ST Array 10833777 - -
          Affymetrix Rat Gene 1.0 ST Array 10833771 - -
          Affymetrix Rat Gene 1.0 ST Array 10833783 - -
          Affymetrix Rat Gene 1.0 ST Array 10833780 - -
          Affymetrix Rat Gene 1.0 ST Array 10833772 - -
          Affymetrix Rat Gene 1.0 ST Array 10833773 - -
          Affymetrix Rat Gene 1.0 ST Array 10833781 - -
          Affymetrix Rat Gene 1.0 ST Array 10833778 - -
          Affymetrix Rat Gene 1.0 ST Array 10833779 - -
          Affymetrix Rat Gene 1.0 ST Array 10833776 - -
          Affymetrix Rat Gene 1.0 ST Array 10833775 - -
          Affymetrix Rat Gene 1.1 ST Array 10833770 - -
          Rat Genome 230 2.0 Array 1380259_PM_at - -
          Rat Genome 230 2.0 Array 1380259_at - -
          Affymetrix HT Rat Focus Array 1380259_at - -
          Affymetrix HT Rat Focus Array 1380259_PM_at - -
          Affymetrix Rat Clariom S Assay PSR2000013863.rn.1 TC2000001708.rn.1 RTA TC
          Rat Expression Set 230 1380259_at - -
          Rat Expression Set 230 1380259_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10833770 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22294448 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1350240 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline